Anti-CD8A Rabbit Polyclonal Antibody

Supplier: Sino Biological
60001-RP01-200 60001-RP01-400 60001-RP01-100
103660-154EA 351.49 USD
103660-154 103660-158 103660-156
Anti-CD8A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Ferret CD8a/Lyt2 (Catalog#60001-F08H; ABS50091.1; Met1-Glu186). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR