Anti-CD8A Rabbit Monoclonal Antibody [clone: 080]

Supplier: Sino Biological
60001-R080-100 60001-R080-50
103660-152EA 271.4 USD
103660-152 103596-362
Anti-CD8A Rabbit Monoclonal Antibody [clone: 080]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Ferret CD8a (rF CD8a; Catalog#60001-F08H; ABS50091.1; Met1-Glu186).

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 080
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR