Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD8A Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 073]

Supplier: Sino Biological

60001-R073-F-100 60001-R073-F-25
103632-372EA 172.31 USD InStock
103632-372 103630-192
Anti-CD8A Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 073]
Antibodies
Anti-CD8 alpha Antibody (FITC)

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: Clone: 073
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty