Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD8A Mouse Monoclonal Antibody [clone: 3B5H2]

Supplier: Sino Biological

60001-MM02-1 60001-MM02-50 60001-MM02-200 60001-MM02-100
103589-592EA 958.46 USD InStock
103589-592 103596-358 103660-148 103660-146
Anti-CD8A Mouse Monoclonal Antibody [clone: 3B5H2]
Antibodies
Anti-CD8 alpha Antibody

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: Clone: 3B5H2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Ferret

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty