Anti-APLP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51730-T24-50 51730-T24-100 51730-T24-200
103589-588EA 106.62 USD
103589-588 103596-348 103596-350
Anti-APLP1 Rabbit Polyclonal Antibody
Antibodies
Anti-APLP1 Antibody

Type: Primary
Antigen: APLP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR