Anti-Lp-PLA2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51562-RP01-100 51562-RP01-400 51562-RP01-200
103660-820EA 106.62 USD
103660-820 103660-824 103660-822
Anti-Lp-PLA2 Rabbit Polyclonal Antibody
Antibodies
Anti-Lp-PLA2 Antibody

Type: Primary
Antigen: Lp-PLA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR