Anti-JPT1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51359-RP01-100 51359-RP01-200 51359-RP01-400
103660-142EA 270.2 USD
103660-142 103660-818 103660-144
Anti-JPT1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse HN-1 / ARM2 (rM HN-1 / ARM2; Catalog#51359-M07E; NP_032284.1; Met1-Gly154). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: HN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR