Anti-IL16 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51303-T16-50 51303-T16-200 51303-T16-100
103596-326EA 352.03 USD
103596-326 103589-578 103596-324
Anti-IL16 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL16 (Catalog#51303-M07E; NP_034681.2; Ser1205-Ser1322). IL16 specific IgG was purified by Mouse IL16 affinity chromatography.

Type: Primary
Antigen: IL-16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR