Anti-PSMB6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51239-T48-50 51239-T48-200 51239-T48-100
103589-572EA 142.96 USD
103589-572 103596-316 103596-314
Anti-PSMB6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PSMB6 / Proteasome subunit beta type-6 (rM PSMB6 / Proteasome subunit beta type-6; Catalog#51239-M07E; NP_032972.3; Thr34-Pro238). PSMB6 / Proteasome subunit beta type-6 specific IgG was purified by Mouse PSMB6 / Proteasome subunit beta type-6 affinity chromatography.

Type: Primary
Antigen: PSMB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR