Anti-PSMB6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51239-RP01-200 51239-RP01-100 51239-RP01-400
103660-118EA 562.97 USD
103660-118 103660-116 103660-114
Anti-PSMB6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PSMB6/Proteasome subunit beta type-6 (Catalog#51239-M07E; NP_032972.3; Thr34-Pro238). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PSMB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR