Anti-LDHA Rabbit Polyclonal Antibody

Supplier: Sino Biological
51207-T52-50 51207-T52-200 51207-T52-100
103596-300EA 352.03 USD
103596-300 103589-566 103596-298
Anti-LDHA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LDH-A/LDHA (rM LDH-A/LDHA; Catalog#51207-M07E; NP_034829.1; Met1-Phe332). LDH-A/LDHA specific IgG was purified by Mouse LDH-A/LDHA affinity chromatography.

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR