Anti-TSC22D1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51196-T16-50 51196-T16-100 51196-T16-200
103589-564EA 106.62 USD
103589-564 103596-294 103596-296
Anti-TSC22D1 Rabbit Polyclonal Antibody
Antibodies
Anti-TSC22D1 Antibody

Type: Primary
Antigen: TSC22D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR