Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSC22D1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51196-RP01-200 51196-RP01-100 51196-RP01-400
103660-098EA 160.46 USD InStock
103660-098 103660-100 103660-102
Anti-TSC22D1 Rabbit Polyclonal Antibody
Antibodies
Anti-TSC22D1 Antibody

Type: Primary
Antigen: TSC22D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty