Anti-IL-21R Rabbit Polyclonal Antibody

Supplier: Sino Biological

51184-RP01-200 51184-RP01-100 51184-RP01-400
103660-086EA 160.46 USD
103660-086 103660-088 103660-090
Anti-IL-21R Rabbit Polyclonal Antibody
Antibodies
Anti-IL21 Receptor Antibody

Type: Primary
Antigen: IL-21R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR