Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EGFL6 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51182-RP01-100 51182-RP01-200 51182-RP01-400
103660-082EA 106.62 USD InStock
103660-082 103660-080 103660-084
Anti-EGFL6 Rabbit Polyclonal Antibody
Antibodies
Anti-EGFL6 Antibody

Type: Primary
Antigen: EGFL6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty