Anti-CNTF Rabbit Polyclonal Antibody

Supplier: Sino Biological

51181-T24-100 51181-T24-50 51181-T24-200
103596-278EA 160.46 USD
103596-278 103589-556 103596-280
Anti-CNTF Rabbit Polyclonal Antibody
Antibodies
Anti-CNTF Antibody

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR