Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL-36G Rabbit Polyclonal Antibody

Supplier: Sino Biological

51179-T52-100 51179-T52-50 51179-T52-200
103596-268EA 160.46 USD InStock
103596-268 103589-552 103596-270
Anti-IL-36G Rabbit Polyclonal Antibody
Antibodies
Anti-IL36 gamma Antibody

Type: Primary
Antigen: IL-36G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty