Anti-CD22 Rabbit Monoclonal Antibody [clone: 060] PE (Phycoerythrin)

Supplier: Sino Biological
51177-R060-P-100 51177-R060-P-25
103630-186EA 70.7 USD
103630-186 103632-366
Anti-CD22 Rabbit Monoclonal Antibody [clone: 060] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD22 (rM CD22; Catalog#51177-M08H; BAE33829.1; Met1-Arg708) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: Clone: 060
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR