Anti-CD22 Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 060]

Supplier: Sino Biological

51177-R060-A-100 51177-R060-A-25
103632-362EA 204.62 USD
103632-362 103630-182
Anti-CD22 Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 060]
Antibodies
Anti-CD22 Antibody (APC)

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: Clone: 060
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR