Anti-CD22 Rabbit Monoclonal Antibody [clone: 060]

Supplier: Sino Biological

51177-R060-50 51177-R060-100
103596-254EA 128.15 USD
103596-254 103660-062
Anti-CD22 Rabbit Monoclonal Antibody [clone: 060]
Antibodies
Anti-CD22 Antibody

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: Clone: 060
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR