Anti-CHL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51176-RP01-400 51176-RP01-200 51176-RP01-100
103660-054EA 351.49 USD
103660-054 103660-058 103660-056
Anti-CHL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CHL1 / LICAM2 / CALL (rM CHL1 / LICAM2 / CALL; Catalog#51176-M08H; ; Met1-Gln1027). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR