Anti-SERPINA1D Rabbit Polyclonal Antibody

Supplier: Sino Biological

51172-T16-200 51172-T16-50 51172-T16-100
103596-246EA 214.31 USD
103596-246 103589-546 103596-244
Anti-SERPINA1D Rabbit Polyclonal Antibody
Antibodies
Anti-SERPINA1D Antibody

Type: Primary
Antigen: SERPINA1D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR