Anti-SERPINA1D Rabbit Polyclonal Antibody

Supplier: Sino Biological
51172-RP01-100 51172-RP01-200 51172-RP01-400
103660-052EA 562.97 USD
103660-052 103660-050 103660-048
Anti-SERPINA1D Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SERPINA1D (Catalog#51172-M08H; XP_003945711.1; Met1-Lys413). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SERPINA1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR