Anti-YM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51165-T50-100 51165-T50-200 51165-T50-50
103596-240EA 160.46 USD
103596-240 103596-242 103589-544
Anti-YM1 Rabbit Polyclonal Antibody
Antibodies
Anti-YM1 Antibody

Type: Primary
Antigen: YM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR