Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-YM1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51165-RP01-100 51165-RP01-200 51165-RP01-400
103660-042EA 106.62 USD InStock
103660-042 103660-044 103660-046
Anti-YM1 Rabbit Polyclonal Antibody
Antibodies
Anti-YM1 Antibody

Type: Primary
Antigen: YM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty