Anti-CDH13 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51163-T52-100 51163-T52-50 51163-T52-200
103596-238EA 352.03 USD
103596-238 103589-542 103596-236
Anti-CDH13 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CDH13 / Cadherin-13 / H Cadherin (Catalog#51163-M08H; NP_062681.2; Met1-Ala692). CDH13 / Cadherin-13 / H Cadherin specific IgG was purified by Mouse CDH13 / Cadherin-13 / H Cadherin affinity chromatography.

Type: Primary
Antigen: CDH13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR