Anti-Factor H Rabbit Polyclonal Antibody

Supplier: Sino Biological

51160-RP01-400 51160-RP01-200 51160-RP01-100
103660-816EA 214.31 USD
103660-816 103660-814 103660-812
Anti-Factor H Rabbit Polyclonal Antibody
Antibodies
Anti-Factor H Antibody

Type: Primary
Antigen: Factor H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR