Anti-REG3B Rabbit Monoclonal Antibody [clone: 065]

Supplier: Sino Biological
51153-R065-100 51153-R065-50
103660-810EA 271.4 USD
103660-810 103596-230
Anti-REG3B Rabbit Monoclonal Antibody [clone: 065]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse REG3B (rM REG3B; Catalog#51153-M08H; NP_035166.1; Met1-Gly175).

Type: Primary
Antigen: REG3B
Clonality: Monoclonal
Clone: 065
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR