Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-REG2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51151-T16-200 51151-T16-50 51151-T16-100
103596-224EA 214.31 USD InStock
103596-224 103589-532 103596-222
Anti-REG2 Rabbit Polyclonal Antibody
Antibodies
Anti-REG2 Antibody

Type: Primary
Antigen: REG2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty