Anti-REG2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51151-RP01-200 51151-RP01-400 51151-RP01-100
103660-032EA 270.2 USD
103660-032 103660-030 103660-034
Anti-REG2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Reg2 / Reg-2 (Catalog#51151-M08H; NP_033069.1; Met1-Ala173). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: REG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR