Anti-REG2 Rabbit Monoclonal Antibody [clone: 014]

Supplier: Sino Biological

51151-R014-50 51151-R014-100
103596-220EA 128.15 USD
103596-220 103660-028
Anti-REG2 Rabbit Monoclonal Antibody [clone: 014]
Antibodies
Anti-REG2 Antibody

Type: Primary
Antigen: REG2
Clonality: Monoclonal
Clone: Clone: 014
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR