Anti-CRELD1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51149-RP01-200 51149-RP01-100 51149-RP01-400
103660-022EA 351.49 USD
103660-022 103660-026 103660-024
Anti-CRELD1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CRELD1 (rM CRELD1; Catalog#51149-M08H; NP_598691.1; Met1-Glu362). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CRELD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR