Anti-CRELD2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51148-T16-200 51148-T16-50 51148-T16-100
103596-214EA 214.31 USD
103596-214 103589-528 103596-212
Anti-CRELD2 Rabbit Polyclonal Antibody
Antibodies
Anti-CRELD2 Antibody

Type: Primary
Antigen: CRELD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR