Anti-CNPY3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51146-T16-50 51146-T16-100 51146-T16-200
103596-206EA 270.87 USD
103596-206 103596-208 103589-526
Anti-CNPY3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CNPY3 / TNRC5 (rM CNPY3 / TNRC5; Catalog#51146-M08H; NP_082341.1; Met1-Pro272). CNPY3 / TNRC5 specific IgG was purified by Mouse CNPY3 / TNRC5 affinity chromatography.

Type: Primary
Antigen: CNPY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR