Anti-IL1RL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51143-T48-50 51143-T48-100 51143-T48-200
103596-202EA 270.87 USD
103596-202 103596-204 103589-524
Anti-IL1RL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL1RL1 / IL‑1 R4 (rM IL1RL1 / IL‑1 R4; Catalog#51143-M08H; NP_001020773.1; Met1-Arg332). IL1RL1 / IL‑1 R4 specific IgG was purified by Mouse IL1RL1 / IL‑1 R4 affinity chromatography.

Type: Primary
Antigen: ST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR