Anti-CCL22 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51135-RP01-100 51135-RP01-200 51135-RP01-400
103659-990EA 351.49 USD
103659-990 103659-992 103659-994
Anti-CCL22 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CCL22/MDC (rM CCL22/MDC; Catalog#51135-M07E; O88430; Gly26-Ser92). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: MDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR