Anti-FGFR2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51128-T16-100 51128-T16-200 51128-T16-50
103589-518EA 142.96 USD
103589-518 103596-190 103596-192
Anti-FGFR2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FGFR2/CD332 (Catalog#51128-M08H; AAA39377.1; Met1-Glu263). FGFR2/CD332 specific IgG was purified by Mouse FGFR2/CD332 affinity chromatography.

Type: Primary
Antigen: FGFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR