Anti-FSTL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51127-RP01-400 51127-RP01-200 51127-RP01-100
103659-984EA 351.49 USD
103659-984 103659-986 103659-988
Anti-FSTL1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FSTL1 (Catalog#51127-M08H; NP_032073.2; Met1-Ile306). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR