Anti-IFNA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51126-T60-200 51126-T60-100 51126-T60-50
103596-180EA 214.31 USD
103596-180 103596-178 103589-512
Anti-IFNA5 Rabbit Polyclonal Antibody
Antibodies
Anti-interferon-alpha Antibody

Type: Primary
Antigen: IFNA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR