Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFNA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51126-RP01-400 51126-RP01-100 51126-RP01-200
103659-982EA 214.31 USD InStock
103659-982 103659-980 103659-978
Anti-IFNA5 Rabbit Polyclonal Antibody
Antibodies
Anti-interferon-alpha Antibody

Type: Primary
Antigen: IFNA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty