Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51124-T44-100 51124-T44-200 51124-T44-50
103596-174EA 160.46 USD InStock
103596-174 103596-176 103589-510
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antibodies
Anti-IGFBP2 Antibody

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty