Anti-IGFBP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51124-RP01-100 51124-RP01-400 51124-RP01-200
103659-972EA 351.49 USD
103659-972 103659-974 103659-976
Anti-IGFBP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, human cell-derived, recombinant Human EGFR / ErbB / HER1. Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IGFBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR