Anti-IGFBP2 Rabbit Monoclonal Antibody [clone: 106]

Supplier: Sino Biological
51124-R106-50 51124-R106-100
103596-172EA 150.18 USD
103596-172 103659-970
Anti-IGFBP2 Rabbit Monoclonal Antibody [clone: 106]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IGFBP-2 / IGFBP2 (rM IGFBP-2 / IGFBP2; Catalog#51124-M08H; NP_032368.2; Met1-Gln305).

Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 106
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR