Anti-EphB6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51123-T08-50 51123-T08-200 51123-T08-100
103589-508EA 142.96 USD
103589-508 103596-170 103596-168
Anti-EphB6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EphB6 / Eph Receptor B6 (rM EphB6 / Eph Receptor B6; Catalog#51123-M08H; O08644-1; Met1-Ser587). EphB6 / Eph Receptor B6 specific IgG was purified by Mouse EphB6 / Eph Receptor B6 affinity chromatography.

Type: Primary
Antigen: EphB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR