Anti-M-CSF Rabbit Polyclonal Antibody

Supplier: Sino Biological

51112-T16-50 51112-T16-200 51112-T16-100
103589-504EA 106.62 USD
103589-504 103596-162 103596-160
Anti-M-CSF Rabbit Polyclonal Antibody
Antibodies
Anti-M-CSF Antibody

Type: Primary
Antigen: M-CSF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR