Anti-CDCP1 Rabbit Monoclonal Antibody [clone: 040]

Supplier: Sino Biological
51110-R040-100 51110-R040-50
103659-950EA 271.4 USD
103659-950 103596-146
Anti-CDCP1 Rabbit Monoclonal Antibody [clone: 040]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CDCP1 (rM CDCP1; Catalog#51110-M08H; Q5U462; Met1-Leu666).

Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 040
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR