Anti-IL17RB Rabbit Polyclonal Antibody

Supplier: Sino Biological
51109-RP01-400 51109-RP01-200 51109-RP01-100
103659-942EA 270.2 USD
103659-942 103659-944 103659-946
Anti-IL17RB Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL17BR/IL17RB/IL-17 Receptor B (Catalog#51109-M08H; Q9JIP3-1; Met1-Gly286). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: IL17RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR