Anti-IL27 Rabbit Polyclonal Antibody

Supplier: Sino Biological
51107-T16-100 51107-T16-200 51107-T16-50
103589-494EA 142.96 USD
103589-494 103596-136 103596-134
Anti-IL27 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IL-27 (Catalog#51107-M08H; O35228 and Q8K3I6; Met1-Pro228 and Phe29-Ser234). IL-27 specific IgG was purified by Mouse IL-27 affinity chromatography.

Type: Primary
Antigen: IL-27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR