Anti-NTRK1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

51103-T16-100 51103-T16-50 51103-T16-200
103596-126EA 160.46 USD
103596-126 103589-490 103596-128
Anti-NTRK1 Rabbit Polyclonal Antibody
Antibodies
Anti-TrkA Antibody

Type: Primary
Antigen: NTRK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR