Anti-IL17F Rabbit Polyclonal Antibody

Supplier: Sino Biological
51097-T48-50 51097-T48-100 51097-T48-200
103596-118EA 157.27 USD
103596-118 103589-486 103596-120
Anti-IL17F Rabbit Polyclonal Antibody
Antibodies
Anti-IL17F Antibody

Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR